Learn More
Medchemexpress LLC HY-105182A 100mg , Xanomeline (tartrate) CAS:152854-19-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105182A100MG
Encompass_Preferred
HY-105182A 100mg Xanomeline (tartrate) CAS: 152854-19-8 Xanomeline (LY 246708) is the potent agonist of muscarinic M1/M4 receptor with antipsychotic-like activity. Xanomeline (LY 246708) increases neuronal excitability. Xanomeline (LY 246708) can be used for the research of schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1262