Learn More
Medchemexpress LLC Xanomeline tartrate | 152854-19-8 | 99.9% | 431.50 g/mol | C18H29N3O7S | 5 MG
SDP
Catalog No. 5000242507
Encompass_Preferred
Xanomeline tartrate is the tartrate salt of xanomeline, a muscarinic M1/M4 receptor agonist used in preclinical neuroscience and pharmacology research. It is supplied as a solid and as a DMSO solution for analytical and research applications.
  • Selective muscarinic M1/M4 receptor agonist suitable for receptor pharmacology studies.
  • High purity (LCMS 99.9%) appropriate for analytical and preclinical use.
  • Available in small solid quantities and as a 10 mM DMSO solution for flexible dosing and testing.
  • Tartrate salt form improves solubility and handling compared with the free base.
  • Molecular weight 431.50 g/mol and CAS 152854-19-8 for unambiguous identification.
Catalog No. 50-002-42507 Supplier Medchemexpress LLC Supplier No. HY105182A5MG
Xanomeline tartrate is the tartrate salt of xanomeline, a muscarinic M1/M4 receptor agonist used in preclinical neuroscience and pharmacology research. It is supplied as a solid and as a DMSO solution for analytical and research applications.
  • Selective muscarinic M1/M4 receptor agonist suitable for receptor pharmacology studies.
  • High purity (LCMS 99.9%) appropriate for analytical and preclinical use.
  • Available in small solid quantities and as a 10 mM DMSO solution for flexible dosing and testing.
  • Tartrate salt form improves solubility and handling compared with the free base.
  • Molecular weight 431.50 g/mol and CAS 152854-19-8 for unambiguous identification.