Learn More
Medchemexpress LLC Xanomeline tartrate | 152854-19-8 | MFCD04034716 | 99.9% | 431.50 | C18H29N3O7S | 50 MG
SDP
Catalog No. 5000243704
Encompass_Preferred
Xanomeline tartrate is the tartrate salt of xanomeline, a muscarinic acetylcholine receptor agonist with functional selectivity for M1 and M4 subtypes. It is supplied as a research-grade solid for pharmacology and behavioral studies investigating cholinergic signaling and receptor pharmacology.
  • Muscarinic M1/M4 receptor agonist.
  • Tartrate salt form for improved stability and solubility.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High purity (99.9% by LCMS).
  • Molecular weight 431.50.
Catalog No. 50-002-43704 Supplier Medchemexpress LLC Supplier No. HY105182A50MG
Xanomeline tartrate is the tartrate salt of xanomeline, a muscarinic acetylcholine receptor agonist with functional selectivity for M1 and M4 subtypes. It is supplied as a research-grade solid for pharmacology and behavioral studies investigating cholinergic signaling and receptor pharmacology.
  • Muscarinic M1/M4 receptor agonist.
  • Tartrate salt form for improved stability and solubility.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High purity (99.9% by LCMS).
  • Molecular weight 431.50.