Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanomeline tartrate | 152854-19-8 | MFCD04034716 | 99.9% | 431.50 g·mol⁻1 | C18H29N3O7S | 10 MG
SDP

Catalog No. 5000241829
Encompass_Preferred

Xanomeline tartrate is the tartrate salt of xanomeline, a selective muscarinic M1/M4 receptor agonist supplied as a high-purity research reagent for in vitro pharmacology and preclinical neuronal signaling studies.

  • High purity: 99.9%.
  • Molecular weight 431.50 g·mol⁻1.
  • Chemical formula C18H29N3O7S.
  • CAS number 152854-19-8.
  • Primary target muscarinic M1 and M4 receptors.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-41829 Supplier Medchemexpress LLC Supplier No. HY105182A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanomeline tartrate is the tartrate salt of xanomeline, a selective muscarinic M1/M4 receptor agonist supplied as a high-purity research reagent for in vitro pharmacology and preclinical neuronal signaling studies.

  • High purity: 99.9%.
  • Molecular weight 431.50 g·mol⁻1.
  • Chemical formula C18H29N3O7S.
  • CAS number 152854-19-8.
  • Primary target muscarinic M1 and M4 receptors.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.