Learn More
Medchemexpress LLC Xanomeline (tartrate) | 152854-19-8 | MFCD04034716 | 99.9% | 431.50 g·mol⁻¹ | C18H29N3O7S | 25 MG
SDP
Catalog No. 5000244116
Encompass_Preferred
Xanomeline tartrate is the tartrate salt of xanomeline, a muscarinic acetylcholine receptor agonist with functional selectivity for M1 and M4 subtypes. It is provided as a research-grade analytical reagent for neuropharmacology and receptor profiling applications, with supporting documentation for characterization and handling.
  • Muscarinic M1/M4 receptor agonist suitable for neuropharmacology research.
  • Tartrate salt form to improve compound stability and handling.
  • High purity (LCMS 99.9%) for reliable experimental results.
  • Provided in small, defined milligram quantities for analytical use.
  • Documented molecular formula and molecular weight for characterization.
Catalog No. 50-002-44116 Supplier Medchemexpress LLC Supplier No. HY105182A25MG
Xanomeline tartrate is the tartrate salt of xanomeline, a muscarinic acetylcholine receptor agonist with functional selectivity for M1 and M4 subtypes. It is provided as a research-grade analytical reagent for neuropharmacology and receptor profiling applications, with supporting documentation for characterization and handling.
  • Muscarinic M1/M4 receptor agonist suitable for neuropharmacology research.
  • Tartrate salt form to improve compound stability and handling.
  • High purity (LCMS 99.9%) for reliable experimental results.
  • Provided in small, defined milligram quantities for analytical use.
  • Documented molecular formula and molecular weight for characterization.