Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthan Gum, FCC, 4.2-5.4%, Spectrum™ Chemical
SDP

Catalog No. 18607751 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
12 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-751 100 g Poly Bottle
18-607-752 500 g Poly Bottle
18-607-753 - Poly Bottle
18-607-754 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-751 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. X1001100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

X1001, 11138-66-2

Spectrum™ Chemical Xanthan Gum, FCC is a polysaccharide most often used as a food thickening agent and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 11138-66-2
Molecular Formula C8H14Cl2N2O2
Molecular Weight (g/mol) 241.11
InChI Key VXYWXJXCQSDNHX-UHFFFAOYSA-N
IUPAC Name 2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride
SMILES Cl.Cl.NC1=CC(N)=C(OCCO)C=C1

Specifications

Packaging Poly Bottle
Quantity 100 g
Viscosity To pass test
Percent Purity 4.2 to 5.4%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Xanthan Gum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.