Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthan Gum, NF, 91-108%, Spectrum™ Chemical
SDP

Catalog No. 18607755 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-755 100 g Poly Bottle
18-607-756 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-755 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. XA105100GM
Add to Cart
Add to Cart

XA105, 11138-66-2

Spectrum™ Chemical Xanthan Gum, NF is a natural gum polysaccharide used as a thickener or rheology modifier and emulsion stabilizer. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 11138-66-2
Molecular Formula C8H14Cl2N2O2
Molecular Weight (g/mol) 241.11
InChI Key VXYWXJXCQSDNHX-UHFFFAOYSA-N
IUPAC Name 2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride
SMILES Cl.Cl.NC1=CC(N)=C(OCCO)C=C1

Specifications

Packaging Poly Bottle
Quantity 100 g
Percent Purity 91 to 108%
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.