Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthan Gum (Practical Grade), MP Biomedicals

Catalog No. ICN96002190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN96002190 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN96002190 Supplier MP Biomedicals, Inc Supplier No. 0296002190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 11138-66-2
Molecular Formula C8H14Cl2N2O2
Molecular Weight (g/mol) 241.112
InChI Key VXYWXJXCQSDNHX-UHFFFAOYSA-N
Synonym 2-2,4-diaminophenoxy ethanol dihydrochloride, 2,4-diaminophenoxyethanol dihydrochloride, 2,4-diaminophenoxyethanol hcl, unii-z5y843j6kq, ethanol, 2-2,4-diaminophenoxy-, dihydrochloride, 2,4-diaminophenoxyethanol 2hcl, 2-2,4-diaminophenoxy ethanol hydrochloride, ethanol, 2-2,4-diaminophenoxy-, hydrochloride 1:2, 4-2-hydroxyethoxy-1,3-phenylenediamine dihydrochloride, 2,4-diaminophenoxyethanol hydrochloride
PubChem CID 47932
IUPAC Name 2-(2,4-diaminophenoxy)ethanol;dihydrochloride
SMILES C1=CC(=C(C=C1N)N)OCCO.Cl.Cl

Specifications

Quantity 500 g
Grade Practical
Chemical Name or Material Xanthan Gum
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.