Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthine-13C,15N2 | 1262670-81-4 | >98.0% | 155.09 | C413CH4N215N2O2 | 1 MG
SDP

Catalog No. 5000301751
Encompass_Preferred

Xanthine-13C,15N2 is a stable isotope-labeled xanthine supplied for research use as an internal standard, tracer, or for metabolic and mass spectrometry studies. It carries one 13C and two 15N labels and is provided as a dry powder suitable for analytical workflows.

  • Contains 13C and 15N stable isotope labels.
  • Suitable as an internal standard for LC-MS and tracer studies.
  • High reported purity (≥98.0%).
  • Soluble in water and 1 M NaOH under recommended conditions.
  • Supplied in small milligram pack sizes for analytical applications.

Catalog No. 50-003-01751 Supplier Medchemexpress LLC Supplier No. HYW017389S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthine-13C,15N2 is a stable isotope-labeled xanthine supplied for research use as an internal standard, tracer, or for metabolic and mass spectrometry studies. It carries one 13C and two 15N labels and is provided as a dry powder suitable for analytical workflows.

  • Contains 13C and 15N stable isotope labels.
  • Suitable as an internal standard for LC-MS and tracer studies.
  • High reported purity (≥98.0%).
  • Soluble in water and 1 M NaOH under recommended conditions.
  • Supplied in small milligram pack sizes for analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.