Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthine-13C,15N2 | 1262670-81-4 | MFCD34676312 | >97.0% | 155.09 g·mol⁻1 | C4H4N2O2 (isotopically labeled: 13C, 15N2) | 5 MG
SDP

Catalog No. 5000300462
Encompass_Preferred

Xanthine-13C,15N2 is an isotopically labeled xanthine used as a tracer and internal standard in mass spectrometry, metabolic pathway studies, and other biochemical research. The compound contains one 13C atom and two 15N atoms to enable highly specific tracking and quantification in isotope-dilution assays. Supplied in small masses for analytical workflows, it is intended for research use only.

  • Isotopic labeling: 13C and 15N2 for tracer experiments.
  • Suitable as an internal standard for mass spectrometry workflows.
  • Molecular weight 155.09 g·mol⁻1 and formula C4H4N2O2 (isotopically labeled).
  • Supplied in small quantities appropriate for analytical assays.
  • Recommended storage: powder -20°C for long term, 4°C for shorter term.

Catalog No. 50-003-00462 Supplier Medchemexpress LLC Supplier No. HYW017389S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthine-13C,15N2 is an isotopically labeled xanthine used as a tracer and internal standard in mass spectrometry, metabolic pathway studies, and other biochemical research. The compound contains one 13C atom and two 15N atoms to enable highly specific tracking and quantification in isotope-dilution assays. Supplied in small masses for analytical workflows, it is intended for research use only.

  • Isotopic labeling: 13C and 15N2 for tracer experiments.
  • Suitable as an internal standard for mass spectrometry workflows.
  • Molecular weight 155.09 g·mol⁻1 and formula C4H4N2O2 (isotopically labeled).
  • Supplied in small quantities appropriate for analytical assays.
  • Recommended storage: powder -20°C for long term, 4°C for shorter term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.