Learn More
Medchemexpress LLC Xanthine-15N2 | 161770-53-2 | 99.4% | 154.1 g/mol | C5H4(15N)2O2 | 1 MG
SDP
Catalog No. 5000300165
Encompass_Preferred
Xanthine-15N2 is a 15N2 isotopically labeled xanthine provided as a 1 mg powder for research use. It is designed as an internal standard or tracer for mass spectrometry and biochemical assays, offering consistent isotopic labeling and analytical performance.
  • Stable isotope-labeled compound for analytical and tracer studies.
  • High chemical purity (99.4%) suitable for sensitive assays.
  • Small 1 mg powder format for standards and method development.
  • Soluble at 10 mM in DMSO, compatible with common solvents.
  • Long-term storage stability at -20°C (3 years) and 4°C (2 years).
  • For research use only; not for human or clinical use.
Catalog No. 50-003-00165 Supplier Medchemexpress LLC Supplier No. HYW017389S11MG
Xanthine-15N2 is a 15N2 isotopically labeled xanthine provided as a 1 mg powder for research use. It is designed as an internal standard or tracer for mass spectrometry and biochemical assays, offering consistent isotopic labeling and analytical performance.
  • Stable isotope-labeled compound for analytical and tracer studies.
  • High chemical purity (99.4%) suitable for sensitive assays.
  • Small 1 mg powder format for standards and method development.
  • Soluble at 10 mM in DMSO, compatible with common solvents.
  • Long-term storage stability at -20°C (3 years) and 4°C (2 years).
  • For research use only; not for human or clinical use.