Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthine | 161770-53-2 | 99.4% | 154.10 g/mol | C5H4N2(15N)2O2 | 5 MG
SDP

Catalog No. 5000300166
Encompass_Preferred

Xanthine-15N2 is a stable isotope-labeled xanthine supplied as a high-purity analytical standard for mass spectrometry, tracer studies, and metabolic pathway research. The 15N2 labeling provides mass separation from endogenous xanthine, enabling accurate quantitation and recovery correction in LC-MS workflows. Recommended storage and handling preserve isotopic integrity for analytical use.

  • High isotopic enrichment (15N) for accurate mass spectrometry quantification.
  • High purity suitable for analytical and research applications.
  • Molecular weight 154.10 g/mol and formula C5H4N2(15N)2O2 for exact mass calculations.
  • Available in small, pre-weighed packs appropriate for trace-level assays.
  • Stable when stored under recommended conditions to maintain integrity.

Catalog No. 50-003-00166 Supplier Medchemexpress LLC Supplier No. HYW017389S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthine-15N2 is a stable isotope-labeled xanthine supplied as a high-purity analytical standard for mass spectrometry, tracer studies, and metabolic pathway research. The 15N2 labeling provides mass separation from endogenous xanthine, enabling accurate quantitation and recovery correction in LC-MS workflows. Recommended storage and handling preserve isotopic integrity for analytical use.

  • High isotopic enrichment (15N) for accurate mass spectrometry quantification.
  • High purity suitable for analytical and research applications.
  • Molecular weight 154.10 g/mol and formula C5H4N2(15N)2O2 for exact mass calculations.
  • Available in small, pre-weighed packs appropriate for trace-level assays.
  • Stable when stored under recommended conditions to maintain integrity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.