Learn More
Medchemexpress LLC HY-W017389 1g Medchemexpress, Xanthine CAS:69-89-6 Purity:>98%
SDP
Catalog No. 501934630
Encompass_Preferred
Medchemexpress, HY-W017389 1g Xanthine CAS:69-89-6 Xanthine is a purine base found in most human body tissues and fluids and in other organisms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4630 Supplier Medchemexpress LLC Supplier No. HYW0173891G
Medchemexpress, HY-W017389 1g Xanthine CAS:69-89-6 Xanthine is a purine base found in most human body tissues and fluids and in other organisms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.