Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthine | 69-89-6 | 99.1% | 152.11 g/mol | C5H4N4O2 | 50 MG
SDP

Catalog No. 5000301374
Encompass_Preferred

Xanthine is supplied as an analytical reference standard of the purine base xanthine (CAS 69-89-6). It is provided as a solid reference material for research and analytical applications, including method development, calibration, and quality control. The material is offered in small laboratory pack sizes and has a high reported purity.

  • High purity suitable for analytical applications (99.09%).
  • Designed for method development, calibration, and quality control.
  • Supplied as a stable solid reference material for easy handling and storage.
  • Available in small laboratory pack sizes for flexible use.
  • Chemical identity confirmed: C5H4N4O2, molecular weight 152.11 g/mol, CAS 69-89-6.

Catalog No. 50-003-01374 Supplier Medchemexpress LLC Supplier No. HYW017389R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthine is supplied as an analytical reference standard of the purine base xanthine (CAS 69-89-6). It is provided as a solid reference material for research and analytical applications, including method development, calibration, and quality control. The material is offered in small laboratory pack sizes and has a high reported purity.

  • High purity suitable for analytical applications (99.09%).
  • Designed for method development, calibration, and quality control.
  • Supplied as a stable solid reference material for easy handling and storage.
  • Available in small laboratory pack sizes for flexible use.
  • Chemical identity confirmed: C5H4N4O2, molecular weight 152.11 g/mol, CAS 69-89-6.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.