Learn More
Medchemexpress LLC Xanthine | 69-89-6 | MFCD00078453 | 100.0% | 152.11 g/mol | C5H4N4O2 | 50 G
SDP
Catalog No. 5000300588
Encompass_Preferred
Xanthine is a purine base and endogenous metabolite used as a biochemical reagent and reference in research. It participates in purine catabolism and is applied in enzymology, metabolic studies, and analytical assays. The compound is typically supplied as a dry powder that requires appropriate storage and handling per supplier guidance.
  • Purine base and endogenous metabolite used in biochemical research.
  • Suitable for enzymatic assays and metabolic pathway studies.
  • Soluble in alkaline aqueous solutions and moderately soluble in water with heating.
  • Stable as a powder when stored refrigerated or frozen following datasheet recommendations.
  • Offered in multiple package sizes to support small-scale and preparative use.
Catalog No. 50-003-00588 Supplier Medchemexpress LLC Supplier No. HYW01738950G
Xanthine is a purine base and endogenous metabolite used as a biochemical reagent and reference in research. It participates in purine catabolism and is applied in enzymology, metabolic studies, and analytical assays. The compound is typically supplied as a dry powder that requires appropriate storage and handling per supplier guidance.
  • Purine base and endogenous metabolite used in biochemical research.
  • Suitable for enzymatic assays and metabolic pathway studies.
  • Soluble in alkaline aqueous solutions and moderately soluble in water with heating.
  • Stable as a powder when stored refrigerated or frozen following datasheet recommendations.
  • Offered in multiple package sizes to support small-scale and preparative use.