Learn More
Medchemexpress LLC Xanthine | 69-89-6 | MFCD00078453 | 98.9% | 152.11 g/mol | C5H4N4O2 | 5 G
SDP
Catalog No. 5000301373
Encompass_Preferred
Xanthine is a purine-derived endogenous metabolite and intermediate in purine degradation, commonly used as a research standard and reference material in biochemical and analytical workflows. The compound is characterized by the chemical formula C5H4N4O2 and a molecular weight of 152.11 g/mol, and is supplied for laboratory use in small, controlled pack sizes.
  • Provides a stable reference for analytical assays and method development.
  • Suitable for biochemical and enzymology studies of purine metabolism.
  • Available in small pack sizes for laboratory use.
  • Store powder at -20°C for long-term stability; shorter storage at 4°C is possible.
  • High purity grades available to support reliable experimental results.
Catalog No. 50-003-01373 Supplier Medchemexpress LLC Supplier No. HYW0173895G
Xanthine is a purine-derived endogenous metabolite and intermediate in purine degradation, commonly used as a research standard and reference material in biochemical and analytical workflows. The compound is characterized by the chemical formula C5H4N4O2 and a molecular weight of 152.11 g/mol, and is supplied for laboratory use in small, controlled pack sizes.
  • Provides a stable reference for analytical assays and method development.
  • Suitable for biochemical and enzymology studies of purine metabolism.
  • Available in small pack sizes for laboratory use.
  • Store powder at -20°C for long-term stability; shorter storage at 4°C is possible.
  • High purity grades available to support reliable experimental results.