Learn More
Medchemexpress LLC Xanthine | 69-89-6 | MFCD00078453 | 99.1% | 152.11 g/mol | C5H4N4O2 | 100 G
SDP
Catalog No. 5000301354
Encompass_Preferred
Xanthine (CAS 69-89-6) is a purine base and endogenous metabolite involved in purine degradation. It is used as a research reagent and reference standard in biochemical and analytical workflows, including metabolic pathway studies and instrument calibration.
  • Used as a research reagent and reference standard for analytical assays.
  • Reference-standard grades available with documented purity for quantitative work.
  • Well-characterized molecular identity: C5H4N4O2, MW 152.11 g/mol.
  • Suitable for use in chromatography and mass spectrometry calibration.
  • Applicable to studies of purine metabolism and enzymatic pathways.
Catalog No. 50-003-01354 Supplier Medchemexpress LLC Supplier No. HYW017389100G
Xanthine (CAS 69-89-6) is a purine base and endogenous metabolite involved in purine degradation. It is used as a research reagent and reference standard in biochemical and analytical workflows, including metabolic pathway studies and instrument calibration.
  • Used as a research reagent and reference standard for analytical assays.
  • Reference-standard grades available with documented purity for quantitative work.
  • Well-characterized molecular identity: C5H4N4O2, MW 152.11 g/mol.
  • Suitable for use in chromatography and mass spectrometry calibration.
  • Applicable to studies of purine metabolism and enzymatic pathways.