Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthine | 69-89-6 | MFCD00078453 | 99.1% | 152.11 g/mol | C5H4N4O2 | 25 G
SDP

Catalog No. 5000302801
Encompass_Preferred

Xanthine (CAS 69-89-6) is a purine-derived endogenous metabolite used as an analytical reference standard and research reagent in studies of purine metabolism, enzymology, and analytical chemistry. Supplied as a solid standard, it is suitable for quantitative analysis, method development, and calibration.

  • High purity (≈99.1%).
  • Molecular formula C5H4N4O2 and molecular weight 152.11 g/mol.
  • Solid form suitable for analytical reference standards and calibration.
  • Used in metabolic and purine pathway studies, and enzymology.
  • Provided in small sealed quantities appropriate for research and analytical use.

Catalog No. 50-003-02801 Supplier Medchemexpress LLC Supplier No. 25GHYW017389
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthine (CAS 69-89-6) is a purine-derived endogenous metabolite used as an analytical reference standard and research reagent in studies of purine metabolism, enzymology, and analytical chemistry. Supplied as a solid standard, it is suitable for quantitative analysis, method development, and calibration.

  • High purity (≈99.1%).
  • Molecular formula C5H4N4O2 and molecular weight 152.11 g/mol.
  • Solid form suitable for analytical reference standards and calibration.
  • Used in metabolic and purine pathway studies, and enzymology.
  • Provided in small sealed quantities appropriate for research and analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.