Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthine | 69-89-6 | MFCD00078453 | 99.1% | 152.11 g/mol | C5H4N4O2 | 25 MG
SDP

Catalog No. 5000303187
Encompass_Preferred

Xanthine (Standard) is an analytical reference material of xanthine intended for research and method development. It is supplied as a solid reference standard for qualitative and quantitative analyses in chromatographic and mass spectrometry workflows.

  • Reference standard for HPLC, GC, and MS method development and calibration.
  • High purity: 99.09% as stated by manufacturer.
  • Molecular formula C5H4N4O2 and molecular weight 152.11 g/mol.
  • Available in small research quantities (25 mg, 50 mg, 100 mg, 250 mg; larger sizes by quote).
  • Supplied as a white to pale yellow powder; store per certificate of analysis.

Catalog No. 50-003-03187 Supplier Medchemexpress LLC Supplier No. 25MGHYW017389R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthine (Standard) is an analytical reference material of xanthine intended for research and method development. It is supplied as a solid reference standard for qualitative and quantitative analyses in chromatographic and mass spectrometry workflows.

  • Reference standard for HPLC, GC, and MS method development and calibration.
  • High purity: 99.09% as stated by manufacturer.
  • Molecular formula C5H4N4O2 and molecular weight 152.11 g/mol.
  • Available in small research quantities (25 mg, 50 mg, 100 mg, 250 mg; larger sizes by quote).
  • Supplied as a white to pale yellow powder; store per certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.