Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthine | 69-89-6 | MFCD00078453 | 99.1% | 152.11 g/mol | C5H4N4O2 | 250 MG
SDP

Catalog No. 5000302736
Encompass_Preferred

Xanthine (Standard) is an analytical reference standard of xanthine (CAS 69-89-6) intended for research and analytical applications. Xanthine is a plant alkaloid and an intermediate in purine degradation. This high-purity solid is provided for use in method development, calibration, and quality control of analytical assays.

  • High purity (99.09%).
  • Molecular formula C5H4N4O2.
  • Molecular weight 152.11 g/mol.
  • Provided as a stable solid suitable for analytical work.
  • Suitable for calibration, method development, and quality control.
  • Available in 250 mg pack size.

Catalog No. 50-003-02736 Supplier Medchemexpress LLC Supplier No. 250MGHYW017389R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthine (Standard) is an analytical reference standard of xanthine (CAS 69-89-6) intended for research and analytical applications. Xanthine is a plant alkaloid and an intermediate in purine degradation. This high-purity solid is provided for use in method development, calibration, and quality control of analytical assays.

  • High purity (99.09%).
  • Molecular formula C5H4N4O2.
  • Molecular weight 152.11 g/mol.
  • Provided as a stable solid suitable for analytical work.
  • Suitable for calibration, method development, and quality control.
  • Available in 250 mg pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.