Learn More
Sigma Aldrich Fine Chemicals Biosciences Xanthine 9910G
Catalog No. 502899827
Encompass
Xanthine is synthesized from guanine in the presence of guanine deaminase. Xanthine oxidase catalyzes the production of xanthine from hypoxanthine and its subsequent breakdown to uric acid. Xanthine derivatives are present in coffee tea and cocoa seeds. Xanthine has been proposed as a lead for the generation of pharmacologically active compounds.
Catalog No. 50-289-9827 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. X737510G
Xanthine is synthesized from guanine in the presence of guanine deaminase. Xanthine oxidase catalyzes the production of xanthine from hypoxanthine and its subsequent breakdown to uric acid. Xanthine derivatives are present in coffee tea and cocoa seeds. Xanthine has been proposed as a lead for the generation of pharmacologically active compounds.