Learn More
Cusabio Technology LLC xanthine dehydrogenase
Catalog No. 5000051374
Encompass_Preferred
The Mouse Xanthine Oxidase (XO) ELISA Kit is designed to detect and quantify XDH (xanthine dehydrogenase/oxidase) in serum plasma and tissue homogenates with high sensitivity and precision This sandwich ELISA-based assay offers a broad detection range of 7 8 pg/mL to 500 pg/mL and a low detection limit of 1 95 pg/mL ensuring accurate measurement of XDH levels in diverse biological samples The quantitative analysis is completed within 1 5 hours using a sample volume of 50 100 L with absorbance readings taken at 450 nm Ideal for research applications this kit provides reliable results for studying XDH-related metabolic pathways oxidative stress mechanisms or disease biomarkers in mouse models
Catalog No. 50-000-51374 Supplier Cusabio Technology LLC Supplier No. CSBEL026189MO
The Mouse Xanthine Oxidase (XO) ELISA Kit is designed to detect and quantify XDH (xanthine dehydrogenase/oxidase) in serum plasma and tissue homogenates with high sensitivity and precision This sandwich ELISA-based assay offers a broad detection range of 7 8 pg/mL to 500 pg/mL and a low detection limit of 1 95 pg/mL ensuring accurate measurement of XDH levels in diverse biological samples The quantitative analysis is completed within 1 5 hours using a sample volume of 50 100 L with absorbance readings taken at 450 nm Ideal for research applications this kit provides reliable results for studying XDH-related metabolic pathways oxidative stress mechanisms or disease biomarkers in mouse models