Learn More
MOLECULAR DEPOT LLC Xanthine Oxidase - 100 U
Catalog No. 5000013596
Encompass_Preferred
Xanthine Oxidase Grade II (Lyophilized Powder) consists of a highly purified native Xanthine oxidase supplied as a lyophilized form Xanthine oxidase uses molybdenum as cofactor and plays a central role in the metabolism of nucleotides It is extensively used in enzymatic assays for the determination of inorganic phosphorus 5-nucleotidase (5-NT) and adenosine deaminase (ADA) For in vitro research use only
Catalog No. 50-000-13596 Supplier MOLECULAR DEPOT LLC Supplier No. P2010006
Xanthine Oxidase Grade II (Lyophilized Powder) consists of a highly purified native Xanthine oxidase supplied as a lyophilized form Xanthine oxidase uses molybdenum as cofactor and plays a central role in the metabolism of nucleotides It is extensively used in enzymatic assays for the determination of inorganic phosphorus 5-nucleotidase (5-NT) and adenosine deaminase (ADA) For in vitro research use only