Learn More
Medchemexpress LLC Xanthine oxidase, bovine milk | 9002-17-9 | 100 U
SDP
Catalog No. 5000397427
Encompass_Preferred
Xanthine oxidase from bovine milk is a xanthine oxidoreductase enzyme that generates reactive oxygen species (ROS), catalyzes the oxidation of hypoxanthine to xanthine, and further catalyzes the oxidation of xanthine to uric acid.
  • This enzyme rapidly and completely depletes hypoxanthine in human erythrocytes.
  • It inhibits the growth of Plasmodium falciparum in human erythrocytes in vitro.
  • It reduces the viability of postnatal cerebellar granule neurons obtained from 8-day old Sprague-Dawley rat pups.
Catalog No. 50-003-97427 Supplier Medchemexpress LLC Supplier No. HYP2755A100U
Xanthine oxidase from bovine milk is a xanthine oxidoreductase enzyme that generates reactive oxygen species (ROS), catalyzes the oxidation of hypoxanthine to xanthine, and further catalyzes the oxidation of xanthine to uric acid.
  • This enzyme rapidly and completely depletes hypoxanthine in human erythrocytes.
  • It inhibits the growth of Plasmodium falciparum in human erythrocytes in vitro.
  • It reduces the viability of postnatal cerebellar granule neurons obtained from 8-day old Sprague-Dawley rat pups.