Learn More
Medchemexpress LLC Xanthine oxidase, bovine milk | 9002-17-9 | 283 kDa | 5 U
SDP
Catalog No. 5000397450
Encompass_Preferred
Xanthine oxidase, bovine milk is an oxidoreductase enzyme that generates reactive oxygen species (ROS). It catalyzes the oxidation of hypoxanthine to xanthine, and further catalyzes the oxidation of xanthine to uric acid. This product is for research use only.
  • Generates reactive oxygen species
  • Catalyzes oxidation of hypoxanthine to xanthine
  • Catalyzes oxidation of xanthine to uric acid
  • Depletes hypoxanthine in human erythrocytes
  • Inhibits growth of Plasmodium falciparum in human erythrocytes
  • Reduces viability of postnatal cerebellar granule neurons
Catalog No. 50-003-97450 Supplier Medchemexpress LLC Supplier No. HYP2755A5U
Xanthine oxidase, bovine milk is an oxidoreductase enzyme that generates reactive oxygen species (ROS). It catalyzes the oxidation of hypoxanthine to xanthine, and further catalyzes the oxidation of xanthine to uric acid. This product is for research use only.
  • Generates reactive oxygen species
  • Catalyzes oxidation of hypoxanthine to xanthine
  • Catalyzes oxidation of xanthine to uric acid
  • Depletes hypoxanthine in human erythrocytes
  • Inhibits growth of Plasmodium falciparum in human erythrocytes
  • Reduces viability of postnatal cerebellar granule neurons