Learn More
Cayman Chemical Xanthine Oxidase Fluorometric
SDP
Catalog No. 502886472
Encompass_Preferred
XO catalyzes the hydroxylation of hypoxanthine to xanthine and then further catalyzes the oxidation of xanthine to uric acid. When oxidizing NADH, XO generates superoxide, a powerful ROS. XO is found predominantly in the liver and intestine. However, levels of circulating XO can increase dramatically with disease. Cayman’s XO Assay provides a simple and accurate method for quantifying xanthine oxidase activity. The assay is based on a multistep enzymatic reaction whose end product resorufin, a highly fluorescent compound, can be EAsily analyzed using an excitation wavelength between 520-550 nm and an emission wavelength between 585-595 nm.
Catalog No. 50-288-6472 Supplier Cayman Chemical Supplier No. 1001089596 WELL
XO catalyzes the hydroxylation of hypoxanthine to xanthine and then further catalyzes the oxidation of xanthine to uric acid. When oxidizing NADH, XO generates superoxide, a powerful ROS. XO is found predominantly in the liver and intestine. However, levels of circulating XO can increase dramatically with disease. Cayman’s XO Assay provides a simple and accurate method for quantifying xanthine oxidase activity. The assay is based on a multistep enzymatic reaction whose end product resorufin, a highly fluorescent compound, can be EAsily analyzed using an excitation wavelength between 520-550 nm and an emission wavelength between 585-595 nm.