Learn More
Sigma Aldrich Fine Chemicals Biosciences Xanthine Oxidase from bovi
Catalog No. 502903179
Encompass
Xanthine Oxidase (XOD) is a metal flavoprotein. It has flavin adenine dinucleotide (fAD) molybdenum and iron in the ratio 228. This homodimer has a molecule weight of 290kDa. It is a member of the molybdenum-protein family. This enzyme consists of two separated substrate-binding sites.
Catalog No. 50-290-3179 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. X187550UN
Xanthine Oxidase (XOD) is a metal flavoprotein. It has flavin adenine dinucleotide (fAD) molybdenum and iron in the ratio 228. This homodimer has a molecule weight of 290kDa. It is a member of the molybdenum-protein family. This enzyme consists of two separated substrate-binding sites.