Learn More
TARGETMOL CHEMICALS INC XANTHINE OXIDOREDUCTASE- 5MG
SDP
Catalog No. 502579243
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Xanthine oxidoreductase-IN-4 is an orally active xanthine oxidoreductase (XOR) inhibitor that inhibits XOR with an IC50 value of 29.3 nM.Xanthine oxidoreductase-IN-4 can be used to study hyperuricemia. purity: 98%
Catalog No. 50-257-9243 Supplier TARGETMOL CHEMICALS INC Supplier No. T720785MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Xanthine oxidoreductase-IN-4 is an orally active xanthine oxidoreductase (XOR) inhibitor that inhibits XOR with an IC50 value of 29.3 nM.Xanthine oxidoreductase-IN-4 can be used to study hyperuricemia. purity: 98%