Learn More
TARGETMOL CHEMICALS INC XANTHINE OXIDOREDUCTASE- 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T721665MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Xanthine oxidoreductase-IN-5 is an orally active xanthine oxidoreductase (XOR) inhibitor (IC50 55 nM).Xanthine oxidoreductase-IN-5 may be used in studies of acute hyperuricemia. purity: 99%
Catalog No. 50-257-9236