Learn More
TARGETMOL CHEMICALS INC Xanthinol Nicotinate 200MG
SDP
Catalog No. 502877627
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Xanthinol Nicotinate (Complamin) is a potent vasodilator that can easily enter the cell and causes an increase in glucose metabolism resulting in an increased energy. Purity 99.72%
Catalog No. 50-287-7627 Supplier TARGETMOL CHEMICALS INC Supplier No. T22459200MG
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Xanthinol Nicotinate (Complamin) is a potent vasodilator that can easily enter the cell and causes an increase in glucose metabolism resulting in an increased energy. Purity 99.72%