Learn More
Medchemexpress LLC Xanthinol Nicotinate | 437-74-1 | 99.9% | 250 MG
SDP
Catalog No. 5000414494
Encompass_Preferred
Xanthinol Nicotinate is a vasodilator that acts directly on the smooth muscle of small arteries and capillaries. It expands blood vessels, improves blood rheology, and reduces peripheral vascular resistance.
  • Inhibits human umbilical artery smooth muscle cell proliferation in a dose-dependent manner.
  • Decreases PDGFR mRNA and PDGFR-β levels on the cell membranes of HUASMCs.
  • Modifies tumor pO2 in vivo.
  • Radiosensitizes tumors.
Catalog No. 50-004-14494 Supplier Medchemexpress LLC Supplier No. 250MGHYB1815
Xanthinol Nicotinate is a vasodilator that acts directly on the smooth muscle of small arteries and capillaries. It expands blood vessels, improves blood rheology, and reduces peripheral vascular resistance.
  • Inhibits human umbilical artery smooth muscle cell proliferation in a dose-dependent manner.
  • Decreases PDGFR mRNA and PDGFR-β levels on the cell membranes of HUASMCs.
  • Modifies tumor pO2 in vivo.
  • Radiosensitizes tumors.