Learn More
Medchemexpress LLC HY-B1815 50mg Medchemexpress, Xanthinol Nicotinate CAS:437-74-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB181550MG
Encompass_Preferred
Medchemexpress, HY-B1815 50mg Xanthinol Nicotinate CAS:437-74-1 Xanthinol Nicotinate (Xanthinol Niacinate), a vasodilator, can act directly on the smooth muscle of small arteries and capillaries. Xanthinol Nicotinate expands blood vessels, improves blood rheology and reduces peripheral vascular resistance[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1231