Learn More
Medchemexpress LLC HY-111588 1mg Medchemexpress, Xanthoangelol CAS:62949-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115881MG
Encompass_Preferred
Medchemexpress, HY-111588 1mg Xanthoangelol CAS:62949-76-2 Xanthoangelol, extracted from Angelica keiskei, suppresses obesity-induced inflammatory responses. Xanthoangelol possesses antibacterial activity. Xanthoangelol inhibits monoamine oxidases. Xanthoangelol induces apoptosis in neuroblastoma and leukemia cells[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0796