Learn More
Apexbio Technology LLC Xanthohumol,10mg CAS# 6754-58-1
SDP
Catalog No. 502240141
Encompass_Preferred
Valosin-containing protein (VCP) inhibitor. Binds to the N domain of VCP, suppressing function and impairing autophagosome maturation. Inhibits growth of a wide variety of human cancer cell lines by inhibiting proliferation and inducing apoptosis. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0141 Supplier Apexbio Technology LLC Supplier No. A863010
Valosin-containing protein (VCP) inhibitor. Binds to the N domain of VCP, suppressing function and impairing autophagosome maturation. Inhibits growth of a wide variety of human cancer cell lines by inhibiting proliferation and inducing apoptosis. Other sizes are also available. Please inqury us for quote.