Learn More
TARGETMOL CHEMICALS INC Xanthohumol 10MG
SDP
Catalog No. 502876842
Encompass_Preferred
Also available in 1 mL, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Xanthohumol, also known as 2', 4, 4'-trihydroxy-6'-methoxy-3'-prenylchalcone or desmethylxanthohumol, is a member of the class of compounds known as 3-prenylated chalcones. It inhibits COX-1 and COX-2 activity and shows chemopreventive effects. Purity 99.18%
Catalog No. 50-287-6842 Supplier TARGETMOL CHEMICALS INC Supplier No. T334210MG
Also available in 1 mL, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Xanthohumol, also known as 2', 4, 4'-trihydroxy-6'-methoxy-3'-prenylchalcone or desmethylxanthohumol, is a member of the class of compounds known as 3-prenylated chalcones. It inhibits COX-1 and COX-2 activity and shows chemopreventive effects. Purity 99.18%