Learn More
Selleck Chemical LLC Xanthohumol
SDP
Supplier:  Selleck Chemical LLC S7889100MG
Encompass_Preferred
Xanthohumol a prenylated chalcone from hop inhibits COX-1 and COX-2 activity and shows chemopreventive effects Xanthohumol inhibits diacylglycerol acyltransferase 1 (DGAT1) and DGAT2 with both IC50 of 40 M Xanthohumol is also a potent antiviral agent against a series of DNA and RNA viruses Xanthohumol induces growth inhibition and apoptosis in cancer cells Phase 1
Catalog No. 50-000-03596