Learn More
Enzo Life Sciences Xanthohumol (5mg). CAS: 569-83-5, 6754-58-1
SDP
Supplier:  Enzo Life Sciences ALX350280M005
Encompass_Preferred
Prenylated chalcone found in beer. Potent inhibitor of diacylglycerol acetyltransferase (DGAT). Inhibits DNA polymerase and induces cell differentiation. Has antiproliferative and cytotoxic effects in human cancer cell lines. Inhibits human P450 enzymes. Induces quinone reductase. Inhibits the expression of HIF-1α and VEGF under hypoxic conditions. Induces apoptosis in mature adipocytes. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml), methanol (10mg/ml) or 100% ethanol; soluble in aqueous buffers up to 20μM. Long Term Storage: +4°C.
Catalog No. 50-200-8826