Learn More
Selleck Chemical LLC Xanthohumol 5mg
SDP
Supplier:  Selleck Chemical LLC S78895MG
Encompass_Preferred
Xanthohumol, a prenylated chalcone from hop, inhibitsCOX-1 andCOX-2 activity and shows chemopreventive effects. Xanthohumol inhibitsdiacylglycerol acyltransferase 1 (DGAT1) andDGAT2 with both IC50 of 40 ?M. Xanthohumol is also a potent antiviral agent against a series of DNA and RNA viruses. Xanthohumol induces growth inhibition andapoptosis in cancer cells. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3320