Learn More
Medchemexpress LLC Xanthohumol | 6754-58-1 | 354.40 | 1 ML
SDP
Catalog No. 5000414834
Encompass_Preferred
Xanthohumol is a principal flavonoid derived from hops, acting as an inhibitor of diacylglycerol acyltransferase (DGAT), COX-1, and COX-2. It exhibits anti-cancer and anti-angiogenic properties. Additionally, Xanthohumol demonstrates antiviral activity against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Inhibits diacylglycerol acyltransferase (DGAT), COX-1, and COX-2.
  • Shows anti-cancer activities.
  • Shows anti-angiogenic activities.
  • Has antiviral activity against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
Catalog No. 50-004-14834 Supplier Medchemexpress LLC Supplier No. HYN106710MM/1ML
Xanthohumol is a principal flavonoid derived from hops, acting as an inhibitor of diacylglycerol acyltransferase (DGAT), COX-1, and COX-2. It exhibits anti-cancer and anti-angiogenic properties. Additionally, Xanthohumol demonstrates antiviral activity against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Inhibits diacylglycerol acyltransferase (DGAT), COX-1, and COX-2.
  • Shows anti-cancer activities.
  • Shows anti-angiogenic activities.
  • Has antiviral activity against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).