Learn More
Medchemexpress LLC Xanthohumol | 6754-58-1 | 354.40 | 100 MG
SDP
Catalog No. 5000415899
Encompass_Preferred
Xanthohumol (Standard) is the analytical standard of Xanthohumol, intended for research and analytical applications. It is a principal flavonoid isolated from hops. The compound acts as an inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2, and exhibits anti-cancer and anti-angiogenic activities. Additionally, Xanthohumol demonstrates antiviral activity against various viruses including bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV). As an analytical standard, it is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Xanthohumol
  • Inhibitor of diacylglycerol acetyltransferase (DGAT)
  • Inhibitor of COX-1 and COX-2
  • Anti-cancer activity
  • Anti-angiogenic activity
  • Antiviral activity against BVDV, rhinovirus, HSV-1, HSV-2, and CMV
  • Used in HPLC, GC, and MS for research
Catalog No. 50-004-15899 Supplier Medchemexpress LLC Supplier No. HYN1067R100MG
Xanthohumol (Standard) is the analytical standard of Xanthohumol, intended for research and analytical applications. It is a principal flavonoid isolated from hops. The compound acts as an inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2, and exhibits anti-cancer and anti-angiogenic activities. Additionally, Xanthohumol demonstrates antiviral activity against various viruses including bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV). As an analytical standard, it is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Xanthohumol
  • Inhibitor of diacylglycerol acetyltransferase (DGAT)
  • Inhibitor of COX-1 and COX-2
  • Anti-cancer activity
  • Anti-angiogenic activity
  • Antiviral activity against BVDV, rhinovirus, HSV-1, HSV-2, and CMV
  • Used in HPLC, GC, and MS for research