Learn More
Medchemexpress LLC Xanthohumol | 6754-58-1 | 99.9% | 354.40 | 5 MG
SDP
Catalog No. 5000393294
Encompass_Preferred
Xanthohumol is a principal flavonoid isolated from hops. It inhibits diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2, and demonstrates anti-cancer, anti-angiogenic, and antiviral activities against viruses like bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Derived from hops
  • Inhibits diacylglycerol acetyltransferase (DGAT)
  • Inhibits COX-1 and COX-2
  • Exhibits anti-cancer activity
  • Exhibits anti-angiogenic activity
  • Possesses antiviral activity
  • Classified as a flavonoid, chalcone, phenol, and polyphenol
Catalog No. 50-003-93294 Supplier Medchemexpress LLC Supplier No. 5MGHYN1067
Xanthohumol is a principal flavonoid isolated from hops. It inhibits diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2, and demonstrates anti-cancer, anti-angiogenic, and antiviral activities against viruses like bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Derived from hops
  • Inhibits diacylglycerol acetyltransferase (DGAT)
  • Inhibits COX-1 and COX-2
  • Exhibits anti-cancer activity
  • Exhibits anti-angiogenic activity
  • Possesses antiviral activity
  • Classified as a flavonoid, chalcone, phenol, and polyphenol