Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthohumol | 6754-58-1 | MFCD00210576 | 99.3% | 354.40 g/mol | C21H22O5 | 10 MG
SDP

Catalog No. 5000295626
Encompass_Preferred

Xanthohumol (Standard) is an analytical reference standard of xanthohumol, a prenylated chalconoid flavonoid isolated from hops, supplied for research and analytical use in qualitative and quantitative assays and method development for HPLC, GC, and MS.

  • Purity 99.30%.
  • Molecular formula C21H22O5.
  • Molecular weight 354.40 g/mol.
  • Supplied as a solid, typically 10 mg.
  • Intended for use as an analytical standard for HPLC, GC, and MS assays.
  • Certificate of analysis provided; follow storage conditions in the CoA.

Catalog No. 50-002-95626 Supplier Medchemexpress LLC Supplier No. HYN1067R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthohumol (Standard) is an analytical reference standard of xanthohumol, a prenylated chalconoid flavonoid isolated from hops, supplied for research and analytical use in qualitative and quantitative assays and method development for HPLC, GC, and MS.

  • Purity 99.30%.
  • Molecular formula C21H22O5.
  • Molecular weight 354.40 g/mol.
  • Supplied as a solid, typically 10 mg.
  • Intended for use as an analytical standard for HPLC, GC, and MS assays.
  • Certificate of analysis provided; follow storage conditions in the CoA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.