Learn More
Medchemexpress LLC Xanthohumol | 6754-58-1 | MFCD00210576 | 99.9% | 354.40 g·mol⁻¹ | C21H22O5 | 10 MG
SDP
Catalog No. 5000296087
Encompass_Preferred
Xanthohumol is a prenylated chalcone flavonoid derived from hops used in research for its enzyme inhibitory and bioactive properties. It is supplied as a purified solid reference compound for biochemical assays, cellular studies, and pharmacology research.
  • High purity (99.9%).
  • Reported inhibitor of DGAT and COX enzymes and shows antiviral and anticancer activity.
  • Well defined molecular formula and molecular weight for analytical use.
  • Suitable for biochemical, cellular, and pharmacology studies at milligram scales.
Catalog No. 50-002-96087 Supplier Medchemexpress LLC Supplier No. HYN106710MG
Xanthohumol is a prenylated chalcone flavonoid derived from hops used in research for its enzyme inhibitory and bioactive properties. It is supplied as a purified solid reference compound for biochemical assays, cellular studies, and pharmacology research.
  • High purity (99.9%).
  • Reported inhibitor of DGAT and COX enzymes and shows antiviral and anticancer activity.
  • Well defined molecular formula and molecular weight for analytical use.
  • Suitable for biochemical, cellular, and pharmacology studies at milligram scales.