Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xanthohumol C | 189299-05-6 | 99.2% | 352.38 | C21H20O5 | 10 MG
SDP

Catalog No. 5000296494
Encompass_Preferred

Xanthohumol C is a prenylchalcone natural product isolated from the hops plant Humulus lupulus. It is used in research and has reported cytotoxic and antiproliferative activity in human cancer cell lines.

  • Chemical formula C21H20O5.
  • Molecular weight 352.38.
  • Purity 99.2%.
  • Reported cytotoxicity against HeLa cells (IC50 12.5 μM).
  • Antiproliferative activity against MCF7 cells (IC50 15.7 μM for 2 days, 6.87 μM for 4 days).
  • Supplied as a 10 MG research quantity.

Catalog No. 50-002-96494 Supplier Medchemexpress LLC Supplier No. HYN1176910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xanthohumol C is a prenylchalcone natural product isolated from the hops plant Humulus lupulus. It is used in research and has reported cytotoxic and antiproliferative activity in human cancer cell lines.

  • Chemical formula C21H20O5.
  • Molecular weight 352.38.
  • Purity 99.2%.
  • Reported cytotoxicity against HeLa cells (IC50 12.5 μM).
  • Antiproliferative activity against MCF7 cells (IC50 15.7 μM for 2 days, 6.87 μM for 4 days).
  • Supplied as a 10 MG research quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.