Learn More
Medchemexpress LLC Xanthohumol D | 274675-25-1 | 370.40 | 1 MG
SDP
Catalog No. 5000415122
Encompass_Preferred
Xanthohumol D, isolated from hops, is an inhibitor of quinone reductase-2 (QR-2) with an IC50 value of 110 μM. It binds to the active site of QR-2 and shows antiproliferative activity on human cancer cell lines in vitro.
  • Inhibitor of quinone reductase-2 (QR-2).
  • Binds to the active site of QR-2.
  • Shows antiproliferative activity on human cancer cell lines in vitro.
Catalog No. 50-004-15122 Supplier Medchemexpress LLC Supplier No. HYN10661MG
Xanthohumol D, isolated from hops, is an inhibitor of quinone reductase-2 (QR-2) with an IC50 value of 110 μM. It binds to the active site of QR-2 and shows antiproliferative activity on human cancer cell lines in vitro.
  • Inhibitor of quinone reductase-2 (QR-2).
  • Binds to the active site of QR-2.
  • Shows antiproliferative activity on human cancer cell lines in vitro.