Learn More
Medchemexpress LLC Xanthohumol D | 274675-25-1 | 370.40 | 5 MG
SDP
Catalog No. 5000416348
Encompass_Preferred
Xanthohumol D, isolated from hops, is an inhibitor of quinone reductase-2 (QR-2) with an IC50 value of 110 μM, and binds to the active site of QR-2. It shows antiproliferative activity on human cancer cell lines in vitro.
  • Inhibitor of quinone reductase-2 (QR-2) with an IC50 value of 110 μM
  • Binds to the active site of QR-2
  • Exhibits antiproliferative activity on human cancer cell lines in vitro
Catalog No. 50-004-16348 Supplier Medchemexpress LLC Supplier No. HYN10665MG
Xanthohumol D, isolated from hops, is an inhibitor of quinone reductase-2 (QR-2) with an IC50 value of 110 μM, and binds to the active site of QR-2. It shows antiproliferative activity on human cancer cell lines in vitro.
  • Inhibitor of quinone reductase-2 (QR-2) with an IC50 value of 110 μM
  • Binds to the active site of QR-2
  • Exhibits antiproliferative activity on human cancer cell lines in vitro