Learn More
Medchemexpress LLC Xanthohumol (Standard) | 6754-58-1 | 210576 | 354.40 | 5 MG
SDP
Catalog No. 5000414772
Encompass_Preferred
Xanthohumol (Standard) is the analytical standard of Xanthohumol, intended for research and analytical applications. Xanthohumol is a principal flavonoid isolated from hops, and it acts as an inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2. It exhibits anti-cancer and anti-angiogenic activities. Additionally, Xanthohumol possesses antiviral activity against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Analytical standard for xanthohumol
  • Inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2
  • Shows anti-cancer and anti-angiogenic activities
  • Antiviral activity against BVDV, rhinovirus, HSV-1, HSV-2, and CMV
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-14772 Supplier Medchemexpress LLC Supplier No. HYN1067R5MG
Xanthohumol (Standard) is the analytical standard of Xanthohumol, intended for research and analytical applications. Xanthohumol is a principal flavonoid isolated from hops, and it acts as an inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2. It exhibits anti-cancer and anti-angiogenic activities. Additionally, Xanthohumol possesses antiviral activity against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Analytical standard for xanthohumol
  • Inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2
  • Shows anti-cancer and anti-angiogenic activities
  • Antiviral activity against BVDV, rhinovirus, HSV-1, HSV-2, and CMV
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS