Learn More
Medchemexpress LLC Xanthohumol (Standard) | 6754-58-1 | MFCD00210576 | 354.40 | 50 MG
SDP
Catalog No. 5000414835
Encompass_Preferred
Xanthohumol (Standard) is the analytical standard of Xanthohumol, intended for research and analytical applications. It is a principal flavonoid isolated from hops, acting as an inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2. It exhibits anti-cancer, anti-angiogenic, and antiviral activities against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Analytical standard for research and analytical applications.
  • Inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2.
  • Exhibits anti-cancer and anti-angiogenic activities.
  • Antiviral activity against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-14835 Supplier Medchemexpress LLC Supplier No. HYN1067R50MG
Xanthohumol (Standard) is the analytical standard of Xanthohumol, intended for research and analytical applications. It is a principal flavonoid isolated from hops, acting as an inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2. It exhibits anti-cancer, anti-angiogenic, and antiviral activities against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Analytical standard for research and analytical applications.
  • Inhibitor of diacylglycerol acetyltransferase (DGAT), COX-1, and COX-2.
  • Exhibits anti-cancer and anti-angiogenic activities.
  • Antiviral activity against bovine viral diarrhea virus (BVDV), rhinovirus, HSV-1, HSV-2, and cytomegalovirus (CMV).
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.