Learn More
AdipoGen Xanthomegnin
SDP
Supplier:  AdipoGen BVT0365C500
Encompass_Preferred
Chemical. CAS 1685-91-2. Formula C30H22O12. MW 574.5. Isolated from Penicillium citreo-viride. Genotoxic mycotoxin. Antitumor compound. Potent iNOS NOSII inhibitor. Interferes with cellular respiratory processes.
Catalog No. 50-256-1970