Learn More
ATCC Synechococcus elongatus
Catalog No. 502380406
Encompass
Whole-genome sequenced cyanobacterium. This product is a xenic culture that also contains the secondary culture Bradyrhizobium sp.; this second culture does not affect the primary culture's growth.
Catalog No. 50-238-0406 Supplier ATCC Supplier No. 33912
Whole-genome sequenced cyanobacterium. This product is a xenic culture that also contains the secondary culture Bradyrhizobium sp.; this second culture does not affect the primary culture's growth.